Skip to Content
Merck
All Photos(1)

Key Documents

00848

Sigma-Aldrich

(1R,4S)-cis-4-Acetoxy-2-cyclopenten-1-ol

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(1S,4R)-cis-4-Hydroxy-2-cyclopentenyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$1,280.00

$1,280.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$1,280.00

About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Beilstein:
4350633
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$1,280.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D −67±2°, c = 2.3% in chloroform

mp

49-51 °C

functional group

ester
hydroxyl

SMILES string

CC(=O)O[C@H]1C[C@@H](O)C=C1

InChI

1S/C7H10O3/c1-5(8)10-7-3-2-6(9)4-7/h2-3,6-7,9H,4H2,1H3/t6-,7+/m0/s1

InChI key

IJDYOKVVRXZCFD-NKWVEPMBSA-N

Application

(1R,4S)-cis-4-Acetoxy-2-cyclopenten-1-ol can be used as a reactant in the total synthesis of Bartlett′s brefeldin intermediate,[1] spinosyn A analogs,[2] (+)-7-deaza-5′-noraristeromycin,[3] (±) strychnine and the Wieland-Gumlich aldehyde.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new approach to (+)-brefeldin A via a nickel-catalyzed coupling reaction of cyclopentenyl acetate and lithium 2-furylborate
Kobayashi Y, et al.
Tetrahedron Letters, 37(34), 6125-6128 (1996)
(+)-7-Deaza-5 `-noraristeromycin as an anti-trypanosomal agent
Seley KL, et al.
Journal of Medicinal Chemistry, 40(4), 622-624 (1997)
Synthesis of novel spinosyn A analogues by Pd-mediated transformations
Tietze LF, et al.
Chemistry?A European Journal , 13(30), 8543-8563 (2007)
Asymmetric Total Syntheses of (-)-and (+)-Strychnine and the Wieland-Gumlich Aldehyde
Knight SD, et al.
Journal of the American Chemical Society, 117(21), 5776-5788 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service