Skip to Content
Merck
All Photos(1)

Key Documents

28-4150

Sigma-Aldrich

Sodium standard solution

suitable for atomic absorption spectrometry, 1000 ppm Na, 1 mg/mL Na

Synonym(s):

Sodium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

form

liquid

availability

available only in Japan

concentration

1 mg/mL Na
1000 ppm Na

suitability

suitable for atomic absorption spectrometry

storage temp.

15-25°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
C M Smart et al.
Anaesthesia, 69(2), 111-117 (2014-01-05)
To maximise the effect of a small fluid load, it is occasionally desirable to bolus manually with multiple depressions of a large-capacity syringe. This is usually achieved by placing the syringe on the side port of a three-way tap. We
Leonard B Kirschner
The Journal of experimental biology, 207(Pt 9), 1439-1452 (2004-03-24)
The emphasis in this review will be on Na(+) absorption across the skin and gills of vertebrates and the gills of crustaceans. However, some recent studies of Cl(-) uptake, especially in crustaceans, will also be described.
Lena L Rosenbaek et al.
The Journal of biological chemistry, 289(19), 13347-13361 (2014-03-29)
The thiazide-sensitive sodium chloride cotransporter, NCC, is the major NaCl transport protein in the distal convoluted tubule (DCT). The transport activity of NCC can be regulated by phosphorylation, but knowledge of modulation of NCC trafficking by phosphorylation is limited. In
J Ashby
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 23(4-5), 507-519 (1985-04-01)
The literature indicates that sodium saccharin is non-reactive to DNA and inactive as a gene mutagen in vitro. At elevated dose levels it is capable of producing structural disturbances in eukaryotic chromosomes in vitro, and it shows intermittent activity as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service