Skip to Content
Merck
All Photos(1)

Key Documents

28-3330

Sigma-Aldrich

Sodium periodate

JIS special grade, 99.5-100.3%

Synonym(s):

Sodium (meta)periodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
99.5-100.3%
grade:
JIS special grade
form:
crystalline
Pricing and availability is not currently available.

grade

JIS special grade

Assay

99.5-100.3%

form

crystalline

reaction suitability

reagent type: oxidant

availability

available only in Japan

pH

3.5-5.5 (25 °C, 107 g/L)

mp

300 °C (dec.) (lit.)

SMILES string

[Na+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

Target Organs

thymus gland

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Millicent Winner et al.
Cancer research, 68(18), 7253-7257 (2008-09-17)
Although chemokine and growth factor receptors are attractive and popular targets for cancer therapeutic intervention, structure-based targeting of the ligands themselves is generally not considered practical. New evidence indicates that a notable exception to this is macrophage migration inhibitory factor
Quyen Cheng et al.
PloS one, 10(5), e0124117-e0124117 (2015-05-06)
Neutrophil recruitment (NR) to sites of sterile inflammation plays a key role in tissue damage and healing potential of lesions characteristic to non-infectious inflammatory diseases. Previous studies suggested significant genetic control of neutrophil survival, function, and migration in inflammatory responses
Carrie E Brubaker et al.
Biomaterials, 31(3), 420-427 (2009-10-09)
There is significant need for effective medical adhesives that function reliably on wet tissue surfaces with minimal inflammatory insult. To address these performance characteristics, we have generated a synthetic adhesive biomaterial inspired by the protein glues of marine mussels. In-vivo
Dmitriy Berillo et al.
Macromolecular bioscience, 12(8), 1090-1099 (2012-06-08)
Macroporous scaffolds composed of chitosan and using oxidized dextran as a crosslinker are produced through cryogelation. Introducing gelatin as a third component into the structure results in the formation of mesopores in the pore walls, which are not seen if
Vaibhav Agarwal et al.
Journal of immunology (Baltimore, Md. : 1950), 191(8), 4235-4245 (2013-09-17)
Streptococcus pneumoniae (pneumococcus) is a major human pathogen, which evolved numerous successful strategies to colonize the host. In this study, we report a novel mechanism of pneumococcal-host interaction, whereby pneumococci use a host complement protein C1q, primarily involved in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service