Skip to Content
Merck
All Photos(1)

Key Documents

28-3020

Sigma-Aldrich

Sodium hydroxide solution

0.5 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

3 mmHg ( 37 °C)

form

liquid

availability

available only in Japan

concentration

0.5 M
1/2 N

density

1.02 g/cm3 at 20 °C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sundaram Chandrasekaran et al.
Chemical Society reviews, 48(15), 4178-4280 (2019-06-18)
In recent years, nanocrystals of metal sulfide materials have attracted scientific research interest for renewable energy applications due to the abundant choice of materials with easily tunable electronic, optical, physical and chemical properties. Metal sulfides are semiconducting compounds where sulfur
Olof Idevall-Hagren et al.
The EMBO journal, 34(17), 2291-2305 (2015-07-24)
The extended synaptotagmins (E-Syts) are ER proteins that act as Ca(2+)-regulated tethers between the ER and the plasma membrane (PM) and have a putative role in lipid transport between the two membranes. Ca(2+) regulation of their tethering function, as well
Yuki Itakura et al.
PloS one, 10(9), e0136660-e0136660 (2015-09-04)
Rhythmic motor patterns underlying many types of locomotion are thought to be produced by central pattern generators (CPGs). Our knowledge of how CPG networks generate motor patterns in complex nervous systems remains incomplete, despite decades of work in a variety
Horacio Cardenas et al.
Epigenetics, 9(11), 1461-1472 (2014-12-04)
A key step in the process of metastasis is the epithelial-to-mesenchymal transition (EMT). We hypothesized that epigenetic mechanisms play a key role in EMT and to test this hypothesis we analyzed global and gene-specific changes in DNA methylation during TGF-β-induced
Joshua S McLane et al.
Biophysical journal, 109(2), 249-264 (2015-07-23)
Mechanical properties of the tumor microenvironment have emerged as key factors in tumor progression. It has been proposed that increased tissue stiffness can transform stromal fibroblasts into carcinoma-associated fibroblasts. However, it is unclear whether the three to five times increase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service