Skip to Content
Merck
All Photos(1)

Key Documents

28-2900

Sigma-Aldrich

Sodium hydride

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH
CAS Number:
Molecular Weight:
24.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
grade:
CP
form:
suspension (in oil)
Pricing and availability is not currently available.

grade

CP

form

suspension (in oil)

reaction suitability

reagent type: reductant

availability

available only in Japan

mp

800 °C (dec.) (lit.)

storage temp.

15-25°C

SMILES string

[Na]

InChI

1S/Na.H

InChI key

MPMYQQHEHYDOCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Direct intercalation into C60 results in the superconducting material (NaH)4C60.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Met. Corr. 1 - Skin Corr. 1A - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuo Xing et al.
Organic & biomolecular chemistry, 13(5), 1515-1520 (2014-12-06)
A novel synthetic route leading to N-glycolyl muramyl dipeptide (MDP), a bacterial glycopeptide of particular interest in studies of nucleotide-binding oligomerization domain-containing protein 2 (NOD2), is described. The synthetic strategy hinges on the alkylation of benzylidene-protected glucal with 2-bromopropionic acid
Hiroshi Fukasawa et al.
ACS chemical neuroscience, 5(7), 525-532 (2014-04-18)
Neuropathic pain is a chronic condition resulting from neuronal damage. Pregabalin, the (S)-isomer of 3-isobutyl-γ-aminobutyric acid (GABA), is widely used to treat neuropathic pain, despite the occurrence of central nervous system (CNS)-related side effects such as dizziness and somnolence. Here
Jie Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 11(31), 3744-3749 (2015-04-29)
Highly regular NaTi2 (PO4 )3 nanocubes with synergistic nanocoatings of rutile TiO2 and carbon are prepared as an electrode material for sodium-ion batteries. It exhibits a high rate and ultralong life performance simultaneously, and a capacity retention of 89.3% after
Dalia S Abdelhamid et al.
Biomaterials, 53, 32-39 (2015-04-22)
Cardiovascular disease initiates with the atherogenic cascade of scavenger receptor- (SR-) mediated oxidized low-density lipoprotein (oxLDL) uptake. Resulting foam cell formation leads to lipid-rich lesions within arteries. We designed amphiphilic macromolecules (AMs) to inhibit these processes by competitively blocking oxLDL
Han Shih et al.
Biomacromolecules, 16(7), 1915-1923 (2015-05-23)
Polymers or hydrogels containing modified cyclodextrin (CD) are highly useful in drug delivery applications, as CD is a cytocompatible amphiphilic molecule that can complex with a variety of hydrophobic drugs. Here, we designed modular photoclick thiol-ene hydrogels from derivatives of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service