Skip to Content
Merck
All Photos(1)

Documents

28-1430

Sigma-Aldrich

Silver standard solution

suitable for atomic absorption spectrometry, 1 mg/mL Ag, 1000 ppm Ag

Synonym(s):

Silver nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL Ag
1000 ppm Ag

suitability

suitable for atomic absorption spectrometry

storage temp.

15-25°C

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigesh Shahare et al.
Toxicology mechanisms and methods, 23(3), 161-167 (2013-01-11)
As the use of silver nanoparticles (AgNPs) is increasing fast in industry, food, medicines, etc., exposure to AgNPs is increasing in quantity day by day. So, it is imperative to know the adverse effects of AgNPs in man. In this
Evaldo Marchi et al.
Growth factors (Chur, Switzerland), 30(5), 304-309 (2012-09-29)
The mechanisms underlying pleural inflammation and pleurodesis are poorly understood. We hypothesized that the cytokines transforming growth factor β (TGFβ1) and vascular endothelial growth factor (VEGF) play a major role in pleurodesis after intrapleural silver nitrate (SN) injection. Forty rabbits
F Gagné et al.
Journal of toxicology and environmental health. Part A, 76(8), 479-490 (2013-06-01)
The increasing application of silver nanoparticles (nAg) in various consumer products has raised concerns regarding toxicological impacts in the environment. It is unclear at present whether the toxicity of nAg is mainly the result of the release of ionic Ag(+)
Takamitsu Miyayama et al.
Toxicology, 305, 20-29 (2013-01-22)
Silver (Ag) possesses antibacterial activity and has been used in wound dressings and deodorant powders worldwide. However, the metabolic behavior and biological roles of Ag in mammals have not been well characterized. In the present study, we exposed human bronchial
Duraisamy Kalpana et al.
Enzyme and microbial technology, 52(3), 151-156 (2013-02-16)
Silver nanoparticles were synthesized by biological method using cultural filtrate of Klebsiella pneumoniae cultured under simulated microgravity and silver nitrate solution as precursor. The nanoparticles exhibited typical plasmon absorption maximum of silver nanoparticles between 405 and 407 nm. Spherical silver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service