Skip to Content
Merck
All Photos(1)

Key Documents

27-0519

Sigma-Aldrich

Rongalit® dihydrate

SAJ first grade, 96.0-103.0%

Synonym(s):

Sodium hydroxymethanesulfinate hydrate, Hydroxymethanesulfinic acid sodium salt, Rongalit®, Sodium formaldehyde sulfoxylate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2SO2Na·xH2O
CAS Number:
Molecular Weight:
118.09 (anhydrous basis)
Beilstein:
3630814
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
96.0-103.0%
grade:
SAJ first grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

96.0-103.0%

form

powder

availability

available only in Japan

pH

9.5-10.5

mp

~120 °C (dec.)

SMILES string

O.[Na+].OCS([O-])=O

InChI

1S/CH4O3S.Na.H2O/c2-1-5(3)4;;/h2H,1H2,(H,3,4);;1H2/q;+1;/p-1

InChI key

ZKODLPLYXXUCHB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Rongalit is a registered trademark of BASF SE

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S V Makarov et al.
Archives of biochemistry and biophysics, 367(2), 289-296 (1999-07-09)
Sodium hydroxymethanesulfinate, (HOCH2SO2Na, HMS) is relatively stable in aqueous alkaline environments, but rapidly decomposes in acidic medium to give a variety of products that include sulfur dioxide. A detailed kinetic and mechanistic study of the decomposition of HMS in slightly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service