Skip to Content
Merck
All Photos(1)

Key Documents

24-5270

Sigma-Aldrich

Potassium phosphate monobasic

SAJ first grade, ≥99.0%

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0%
grade:
SAJ first grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

4.2-4.6 (25 °C, 50 g/L)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Przemyslaw Nogly et al.
IUCrJ, 2(Pt 2), 168-176 (2015-04-14)
Lipidic cubic phases (LCPs) have emerged as successful matrixes for the crystallization of membrane proteins. Moreover, the viscous LCP also provides a highly effective delivery medium for serial femtosecond crystallography (SFX) at X-ray free-electron lasers (XFELs). Here, the adaptation of
Leo Syrjänen et al.
Frontiers in zoology, 12, 19-19 (2015-08-25)
Carbonic anhydrases (CAs, EC 4.2.1.1) are ubiquitous enzymes that catalyze the reversible hydration reaction of carbon dioxide. CAs are present as six structurally divergent enzyme families: α, β, γ, δ, ζ and η. β-CAs have a wide distribution across different
Pamela Gamba et al.
PLoS genetics, 11(6), e1005047-e1005047 (2015-06-26)
Gene expression can be highly heterogeneous in isogenic cell populations. An extreme type of heterogeneity is the so-called bistable or bimodal expression, whereby a cell can differentiate into two alternative expression states. Stochastic fluctuations of protein levels, also referred to
Suvi Vartiainen et al.
Disease models & mechanisms, 7(6), 635-648 (2014-05-09)
A point mutation in the Drosophila gene that codes for the major adult isoform of adenine nuclear translocase (ANT) represents a model for human diseases that are associated with ANT insufficiency [stress-sensitive B(1) (sesB(1))]. We characterized the organismal, bioenergetic and
Jian Zheng et al.
PloS one, 9(4), e94712-e94712 (2014-04-15)
The human organic anion transporting polypeptide 1A2 (OATP1A2) is an important membrane protein that mediates the cellular influx of various substances including drugs. Previous studies have shown that PDZ-domain containing proteins, especially PDZK1 and NHERF1, regulate the function of related

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service