Skip to Content
Merck
All Photos(1)

Documents

24-4110

Sigma-Aldrich

Potassium bisulfate

JIS special grade, ≥98.0%

Synonym(s):

Potassium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHSO4
CAS Number:
Molecular Weight:
136.17
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

214 °C (lit.)

density

2.32 g/mL at 25 °C (lit.)

SMILES string

[K+].OS([O-])(=O)=O

InChI

1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1

InChI key

CHKVPAROMQMJNQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Paananen et al.
Bioresource technology, 193, 97-102 (2015-06-30)
High-alkali low-temperature polysulfide pulping (HALT) was effectively utilised to prevent major polysaccharide losses while maintaining the delignification rate. A yield increase of 6.7 wt% on wood was observed for a HALT pulp compared to a conventionally produced kappa number 60
Antonio Scarano et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 68(5), 964-968 (2010-03-13)
Calcium sulfate (CaS) is a simple, biocompatible material with a very long, safe clinical history in several different fields of medicine. CaS is a rapidly resorbing material that leaves behind a calcium phosphate lattice, which promotes bone regeneration and hemostasis.
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined
Senthaamarai Rogawansamy et al.
International journal of environmental research and public health, 12(6), 6319-6332 (2015-06-05)
Fungal contamination in indoor environments has been associated with adverse health effects for the inhabitants. Remediation of fungal contamination requires removal of the fungi present and modifying the indoor environment to become less favourable to growth.  This may include treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service