Skip to Content
Merck
All Photos(1)

Documents

24-3875

Sigma-Aldrich

Ponceau S solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

availability

available only in Japan

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-8-3-14(11-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,27H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23+,26-25+;;;;

InChI key

KQHKSGRIBYJYFX-WNDMICSESA-J

Looking for similar products? Visit Product Comparison Guide

Suitability

for electrophoresis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jena J Steinle et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 64(3), 325-331 (2009-02-03)
Recent work has suggested that inflammation is a common component of a number of age-related diseases. The hypothesis of the present study was that normal aging of the retina and choroid would increase levels of inducible nitric oxide synthase (iNOS)
Ling Zheng et al.
Proteomics, 9(7), 1869-1882 (2009-03-17)
Retinal ischemia contributes to multiple ocular diseases while aminoguanidine (AMG) treatment significantly inhibits the neuronal and vascular degeneration due to acute retinal ischemia and reperfusion (I/R) injury. In the present study, 2-D DIGE was applied to profile global protein expression
Julia Esbelin et al.
Journal of bacteriology, 191(13), 4419-4426 (2009-04-28)
In the food-borne pathogen Bacillus cereus F4430/73, the production of major virulence factors hemolysin BL (Hbl) and nonhemolytic enterotoxin (Nhe) is regulated through complex mechanisms. The two-component regulatory system ResDE is involved in the activation of hbl and nhe transcription.
S Armando Villalta et al.
Human molecular genetics, 18(3), 482-496 (2008-11-11)
Duchenne muscular dystrophy (DMD) is the most common, lethal, muscle-wasting disease of childhood. Previous investigations have shown that muscle macrophages may play an important role in promoting the pathology in the mdx mouse model of DMD. In the present study
T G van Kooten et al.
Journal of biomedical materials research, 54(3), 385-389 (2001-02-24)
Research of cell-biomaterial interactions is building on knowledge and methods available in cell and molecular biology. Western blotting is one of the options to characterize protein expression in cell populations. Method transfer to biomaterial model systems is not trivial because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service