Skip to Content
Merck
All Photos(1)

Key Documents

24-0880

Sigma-Aldrich

Pentaerythritol

CP

Synonym(s):

2,2-Bis(hydroxymethyl)-1,3-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2OH)4
CAS Number:
Molecular Weight:
136.15
Beilstein:
1679274
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

CP

vapor pressure

<1 mmHg ( 20 °C)

form

solid

availability

available only in Japan

bp

276 °C/30 mmHg (lit.)

mp

253-258 °C (lit.)

SMILES string

OCC(CO)(CO)CO

InChI

1S/C5H12O4/c6-1-5(2-7,3-8)4-9/h6-9H,1-4H2

InChI key

WXZMFSXDPGVJKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jundong Zhu et al.
Journal of biomedical materials research. Part A, 97(4), 498-508 (2011-04-22)
Folate (FA)-conjugated star-shaped copolymer was prepared as a targeted carrier for anticancer drug delivery by ring-opening polymerization of L-lactide using pentaerythritol (PTL) as an initiator, followed by conjugation with methoxy poly(ethylene glycol) (MPEG) and FA-poly(ethylene glycol) (FA-PEG). The resulting amphiphilic
Zhiming Zhang et al.
Organic & biomolecular chemistry, 9(24), 8220-8223 (2011-10-13)
A trimer of cholic acid made by linking three cholic acid molecules with pentaerythritol via click chemistry serves as the center of an artificial metallohydrolase after the complexation of the triazole groups with a Zn(2+) ion. The invertible amphiphilic cavity
Mohamed Touaibia et al.
Chemical communications (Cambridge, England), (4)(4), 380-382 (2007-01-16)
Several oligomannoside clusters having a hundred-fold increase in affinities toward E. coli were synthesized by Cu(I)-catalyzed [1,3]-dipolar cycloadditions using pentaerythritol scaffolds bearing either alkyne or azide functionalities.
Michael C Hacker et al.
Biomacromolecules, 9(6), 1558-1570 (2008-05-17)
In this study, we synthesized and characterized a series of macromers based on poly( N-isopropylacrylamide) that undergo thermally induced physical gelation and, following chemical modification, can be chemically cross-linked. Macromers with number average molecular weights typically ranging from 2000-3500 Da
Guowei Wang et al.
Macromolecular rapid communications, 32(20), 1658-1663 (2011-08-23)
The eight-shaped poly(ethylene oxide) (PEO) is synthesized by a combination of Glaser coupling with ring-opening polymerization (ROP). Firstly, the star-shaped (PEO-OH)(4) is synthesized by ROP of ethylene oxide (EO) using pentaerythritol as an initiator and diphenylmethyl potassium (DPMK) as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service