17-0010
Kaolin
CP
Synonym(s):
Aluminum silicate hydroxide, Bolus, Hydrated aluminum silicate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
~Al2Si2O5(OH)4
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11111803
grade:
CP
form:
solid
Recommended Products
grade
CP
form
solid
availability
available only in Japan
InChI
1S/2Al.O5Si2.2H2O.2O/c;;1-6(2)5-7(3)4;;;;/h;;;2*1H2;;/q2*+1;-2;;;;
InChI key
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ching-Wei Wu et al.
Journal of hazardous materials, 244-245, 412-420 (2013-01-01)
The process of mechanical milling has been found to effectively stabilize heavy metals in municipal solid waste incinerator (MSWI) fly ash, as well as to restrain the evaporation of heavy metals during thermo-treatment. This method is adopted in this study
Kenji Inaba et al.
The journal of trauma and acute care surgery, 74(2), 538-545 (2013-01-29)
The purpose of this study was to evaluate the long-term efficacy and safety of kaolin- and chitosan-based hemostatic agents for hemorrhage control in a 14-day survival, damage-control swine model of Grade IV liver injury. A total of 48 anesthetized pigs
Nicholas R Kunio et al.
American journal of surgery, 205(5), 505-510 (2013-03-19)
The purpose of this study was to compare standard gauze (SG) and advanced hemostatic dressings in use by military personnel in a no-hold model. A randomized, controlled trial was conducted using 36 swine. Animals underwent femoral arteriotomy, followed by 60
B C A Pinheiro et al.
Journal of environmental management, 118, 205-210 (2013-03-05)
This study investigates the incorporation of solid petroleum waste as raw material into a porcelain stoneware tile body, in replacement to natural kaolin material by up to 5 wt.%. Tile formulations containing solid petroleum waste were pressed and fired at 1240 °C
Salvador Cotillas et al.
Water research, 47(5), 1741-1750 (2013-01-29)
In this work, a novel integrated electrochemical process for urban wastewater regeneration is described. The electrochemical cell consists in a Boron Doped Diamond (BDD) or a Dimensionally Stable Anode (DSA) as anode, a Stainless Steel (SS) as cathode and a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service