Skip to Content
Merck
All Photos(1)

Key Documents

15-1300

Sigma-Aldrich

Iron(III) nitrate nonahydrate

JIS special grade, ≥99.0%

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

1.5 (20 °C)

mp

47 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeanet Conradie et al.
Journal of inorganic biochemistry, 100(12), 2069-2073 (2006-11-04)
All low-spin S=1/2 heme-NO complexes feature FeNO angles of about 140 degrees . In contrast, the square-pyramidal [Fe(CN)(4)(NO)](2-) complex features an exactly linear {FeNO}(7) unit. We have sought here to determine a possible, simple molecular orbital (MO) rationale for these
Xianmao Lu et al.
Nano letters, 7(6), 1764-1769 (2007-05-11)
This paper describes a two-step procedure for generating cubic nanocages and nanoframes. In the first step, Au/Ag alloy nanoboxes were synthesized through the galvanic replacement reaction between Ag nanocubes and an aqueous HAuCl4 solution. The second step involved the selective
Marek Basler et al.
Proteomics, 6(23), 6194-6206 (2006-11-30)
Restricting bacterial growth by iron-chelating proteins that reduce iron availability in mucosal secretions and body fluids belongs to basic mechanisms of innate immunity. Most pathogens and commensals thus developed gene regulons responding to iron concentration and encoding iron acquisition systems
A Roberts et al.
Oral microbiology and immunology, 20(3), 147-153 (2005-04-20)
Psychological stress is known to increase the circulating levels of the catecholamine hormones noradrenaline and adrenaline, which have been shown to influence the growth of a large number of bacterial species by acting in a siderophore-like manner or by inducing
F Ndong et al.
International endodontic journal, 45(6), 557-564 (2012-01-25)
To determine the effect of increasing the proportion of zinc on partially stabilized cement (PSC) produced using a one-step sol gel process. A one-step sol-gel process of Portland cement-based PSC with Zn was synthesized by replacing iron nitrate. The crystalline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service