Skip to Content
Merck
All Photos(1)

Key Documents

15-0170

Sigma-Aldrich

Indigo carmine

SAJ first grade, ≥90.0%

Synonym(s):

Acid Blue 74, Indigo-5,5′-disulfonic acid disodium salt, Indigocarmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2Na2O8S2
CAS Number:
Molecular Weight:
466.35
Colour Index Number:
73015
Beilstein:
4103904
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

SAJ first grade

Assay

≥90.0%

form

solid

availability

available only in Japan

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc2NC(\C(=O)c2c1)=C3\Nc4ccc(cc4C3=O)S([O-])(=O)=O

InChI

1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;

InChI key

KHLVKKOJDHCJMG-QDBORUFSSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuneo Oyama
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society, 25 Suppl 1, 7-12 (2013-03-27)
The indication of endoscopic submucosal dissection for Barrett's esophageal adenocarcinoma (BEA) is superficial BEA without lymph node metastasis. The characteristic endoscopic findings of superficial BEA are elevation, depression, and color change. Indigocarmine spreading is useful for the diagnosis of lateral
Kenshi Yao et al.
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society, 25 Suppl 1, 44-54 (2013-02-01)
Because endoscopic submucosal dissection (ESD) of gastric cancer can only be curative if the cancer is detected at an early stage and a precise preoperative diagnosis is made, we reviewed the detection and characterization of early gastric cancer (EGC) using
Christian Gómez-Solís et al.
Journal of hazardous materials, 217-218, 194-199 (2012-04-03)
Indigo carmine and methylene blue dyes in aqueous solution were photodegraded using SiC-TiO(2) catalysts prepared by sol-gel method. After thermal treatment at 450°C, SiC-TiO(2) catalysts prepared in this work showed the presence of SiC and TiO(2) anatase phase. Those compounds
Chaval Sriwong et al.
Journal of environmental sciences (China), 24(3), 464-472 (2012-06-05)
A new method for the preparation of rubber sheet strewn with titanium dioxide particles (TiO2-strewn sheet) is presented. This simple and low cost method is based on the use of TiO2 powder (Degussa P25) being strewn onto the sheet made
Dalel Daâssi et al.
Journal of environmental management, 108, 84-91 (2012-06-05)
Response surface methodology (RSM) was applied to optimize the decolourization of 3 dyes belonging to 3 dye families such as reactive black 5 (diazoic), indigo carmine (indigoid) and aniline blue (anthraquinonic). Crude laccase from Trametes trogii and the laccase-mediator 1-hydroxybenzotriazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service