Skip to Content
Merck
All Photos(1)

Key Documents

12-1820

Sigma-Aldrich

Gum arabic from acacia tree

CP

Synonym(s):

Acacia gum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
13111102
grade:
CP

grade

CP

availability

available only in Japan

dilution

(for analytical testing)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Believed to be a branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts with a mol. wt. of approx. 250,000.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omaima Nasir et al.
Kidney & blood pressure research, 35(5), 365-372 (2012-04-05)
Gum arabic (GA) is a Ca(2+)-, Mg(2+)- and K(+)-rich dietary fiber used for the treatment of patients with chronic kidney disease in Middle Eastern countries. In healthy mice, GA treatment increases creatinine clearance, renal ADH excretion, as well as intestinal
Igor A Schepetkin et al.
International immunopharmacology, 8(10), 1455-1466 (2008-07-04)
Opuntia polyacantha (prickly pear cactus) has been used extensively for its nutritional properties; however, less is known regarding medicinal properties of Opuntia tissues. In the present study, we extracted polysaccharides from O. polyacantha and used size-exclusion chromatography to fractionate the
Badreldin H Ali et al.
PloS one, 8(2), e55242-e55242 (2013-02-06)
Inflammation and oxidative stress are known to be involved in the pathogenesis of chronic kidney disease in humans, and in chronic renal failure (CRF) in rats. The aim of this work was to study the role of inflammation and oxidative
Yadong Yang et al.
Food chemistry, 135(2), 555-561 (2012-08-08)
The interactions between fish gelatin (FG) and gum arabic (GA) in aqueous solutions were investigated by turbidimetry, methylene blue spectrophotometry, zeta potentiometry, dynamic light scattering, protein assay, and state diagram at 40 °C and a total biopolymer concentration (C(T)) of
Shatabhisa Sarkar et al.
Carbohydrate polymers, 90(4), 1685-1694 (2012-09-05)
Spray dried microcapsules of mint oil were prepared using gum Arabic alone and its blends with radiation or enzymatically depolymerized guar gum as wall materials. Microcapsules were evaluated for retention of mint oil during 8-week storage during which qualitative changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service