Skip to Content
Merck
All Photos(1)

Key Documents

12-1605

Sigma-Aldrich

Griess-Romijin nitrite reagent

Synonym(s):

GR nitrite reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
grade:
SAJ special grade
form:
solid

grade

SAJ special grade

form

solid

availability

available only in Japan

storage temp.

15-25°C

Suitability

nitrite detection

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service