Skip to Content
Merck
All Photos(1)

Key Documents

11-0330

Sigma-Aldrich

Florisil® PR

CP, 60-100 mesh

Synonym(s):

Florisil®, Activated magnesium silicate, Magnesium silicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

CP

form

powder

availability

available only in Japan

surface area

289 m2/g

particle size

60-100 mesh

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(16), 6292-6295 (2013-04-12)
Knowledge of the structural properties of mantle phases is critical for understanding the enigmatic seismic features observed in the Earth's lower mantle down to the core-mantle boundary. However, our knowledge of lower mantle phase equilibria at high pressure (P) and
Lei Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 63(10), 2114-2122 (2011-10-08)
Florisil was employed for the sorption of antimony ions from aqueous solutions. A detailed study of the process was performed by varying the sorption time, pH, and temperature. The sorption was found to be fast, equilibrium was reached within 15
Iyad Rashid et al.
International journal of pharmaceutics, 411(1-2), 18-26 (2011-03-23)
A directly compressible excipient has been developed by co-processing starch with magnesium silicate. The foregoing was achieved either by co-precipitation of magnesium silicate onto different types of starch or by dry granulation of maize starch with magnesium silicate. A variety
Sangram H Patil et al.
Journal of chromatography. A, 1216(12), 2307-2319 (2009-02-14)
A multiresidue method is described for simultaneous estimation of 83 pesticides and 12 dioxin-like polychlorinated biphenyls (PCBs) in red and white wines. The samples (20mL wine, acidified with 20 mL 1% HCl) were extracted with 10 mL ethyl acetate (+20
Imran Ali et al.
Chirality, 22(4), 416-424 (2009-07-21)
The contamination of drug residues, including chiral ones, is not acceptable in earth's ecosystem. The dynamicity of enantiomers of thalidomide and its derivatives (3-methyl thalidomide, 3-ethyl thalidomide, and 3-butyl thalidomide) was ascertained at supramolecular level in water-sediment system using solid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service