Skip to Content
Merck
All Photos(1)

Key Documents

09-1480

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt solution

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid disodium salt, EDTA disodium salt, EDTA-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N(CH2CO2Na)CH2CO2H]2
CAS Number:
Molecular Weight:
336.21
Beilstein:
3822669
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

SMILES string

[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2

InChI key

ZGTMUACCHSMWAC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Suitability

for the determination of water hardness 1mL=0.5mg CaO

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suli Huang et al.
PloS one, 10(2), e0117007-e0117007 (2015-02-07)
microRNA (miRNA) plays a role in the pathogenesis of ischemic stroke, and single nucleotide polymorphisms in miRNA genes may contribute to disease susceptibility. However, the effect of miR-146a, miR-196a2, and miR-499 polymorphisms on ischemic stroke susceptibility has been rarely reported.
Haiyan Zhang et al.
Analytica chimica acta, 770, 140-146 (2013-03-19)
A novel method for selective determination of Cr(III) and Cr(VI) in environmental water samples was developed based on target-induced fluorescence quenching of glutathione-stabilized gold nanoclusters (GSH-Au NCs). Fluorescent GSH-Au NCs were synthesized by a one-step approach employing GSH as reducing/protecting
Gervasio A Lamas et al.
JAMA, 309(12), 1241-1250 (2013-03-28)
Chelation therapy with disodium EDTA has been used for more than 50 years to treat atherosclerosis without proof of efficacy. To determine if an EDTA-based chelation regimen reduces cardiovascular events. Double-blind, placebo-controlled, 2 × 2 factorial randomized trial enrolling 1708
G Lima-Oliveira et al.
British journal of biomedical science, 70(1), 6-9 (2013-04-27)
This study assesses the use of different dry K2 (dipotassium) EDTA vacuum tubes and whether or not they might represent a bias in haematological testing. Blood was collected in three dipotassium EDTA vacuum tubes from different manufacturers: Venosafe, Vacuette and
Seung Hyeok Seok et al.
Journal of applied toxicology : JAT, 33(10), 1089-1096 (2013-02-15)
Zinc oxide (ZnO) nanoparticles (NPs) are used in diverse applications ranging from paints and cosmetics to biomedicine and food. Although micron-sized ZnO is a traditional food supplement, ZnO NPs are an unknown public health risk because of their unique physicochemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service