Skip to Content
Merck
All Photos(1)

Key Documents

07-4260

Sigma-Aldrich

N,N-Dimethylaniline

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507140
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.557 (lit.)

pH

7.4 (20 °C, 1.2 g/L)

bp

193-194 °C (lit.)

mp

1.5-2.5 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

CN(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marius Koch et al.
Journal of the American Chemical Society, 134(8), 3729-3736 (2012-01-31)
The fluorescence quenching of 3-cyanoperylene upon electron transfer from N,N-dimethylaniline in three room-temperature ionic liquids (RTILs) and in binary solvent mixtures of identical viscosity has been investigated using steady-state and time-resolved fluorescence spectroscopy. This study was stimulated by previous reports
Kefeng Liu et al.
Dalton transactions (Cambridge, England : 2003), 41(12), 3461-3467 (2012-02-04)
A number of new half-sandwich zirconium(IV) complexes bearing N,N-dimethylaniline-amido ligands with the general formula Cp*ZrCl(2)[ortho-(RNCH(2))(Me(2)N)C(6)H(4)] [R = 2,6-Me(2)C(6)H(3) (1), 2,6-(i)Pr(2)C(6)H(3) (2), (i)Pr (3), (t)Bu (4)] were synthesized by the reaction of Cp*ZrCl(3) with the corresponding ortho-(Me(2)N)C(6)H(4)CH(2)NRLi. All new zirconium complexes
Cai-Rong Zhang et al.
Journal of molecular graphics & modelling, 38, 419-429 (2012-11-03)
The photon to current conversion efficiency of dye-sensitized solar cells (DSCs) can be significantly affected by dye sensitizers. The design of novel dye sensitizers with good performance in DSCs depend on the dye's information about electronic structures and optical properties.
Yong Wang et al.
The journal of physical chemistry. B, 114(8), 2964-2970 (2010-02-12)
This paper addresses the experimentally observed mechanistic differences between the cytochrome P450-catalyzed N-demethylation of substituted N,N-dimethylanilines (DMA) and of N,N-dimethylbenzamides (DMBA). The two reactions of these substrates are initiated by C-H activation of the methyl groups on the nitrogen. Thus
Subashani Maniam et al.
Organic letters, 10(10), 1885-1888 (2008-04-16)
Cyclodextrin [2]rotaxanes have been prepared by coupling dimethylanilines with dicarboxylic acids using DMT-MM, in aqueous solutions of alpha-cyclodextrin, and the example illustrated shows unusual fluorescence emission and other spectroscopic behavior characteristic of the formation of molecular wires in solution, similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service