Skip to Content
Merck
All Photos(1)

Key Documents

05-6000

Sigma-Aldrich

Copper(I) iodide

SAJ first grade, ≥97.0%

Synonym(s):

Cuprous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuI
CAS Number:
Molecular Weight:
190.45
EC Number:
MDL number:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

10 mmHg ( 656 °C)

Assay

≥97.0%

availability

available only in Japan

mp

605 °C (lit.)

density

5.62 g/mL at 25 °C (lit.)

SMILES string

[Cu+].[I-]

InChI

1S/Cu.HI/h;1H/q+1;/p-1

InChI key

LSXDOTMGLUJQCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service