Skip to Content
Merck
All Photos(1)

Key Documents

05-5860

Sigma-Aldrich

Copper(II) chloride dihydrate

SAJ first grade, ≥97.0%

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

>1 (vs air)

Assay

≥97.0%

form

solid

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

pH

3.0-3.8

mp

100 °C (dec.) (lit.)

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Funaki et al.
Organic letters, 14(24), 6186-6189 (2012-12-12)
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.
Shuxia Zhou et al.
Pharmaceutical research, 30(5), 1311-1327 (2013-02-16)
Metal-catalyzed oxidation (MCO) of proteins is of primary concern in the development of biotherapeutics as it represents a prominent degradation pathway with potential undesired biological and biotherapeutic consequences. We developed a fluorogenic derivatization methodology to study the MCO of IgG1
Mónica João Barros Amorim et al.
Environmental pollution (Barking, Essex : 1987), 164, 164-168 (2012-03-01)
Environmental effects of copper nanoparticles are little studied in terrestrial ecosystems. In the present article, the toxicity of copper nanoparticles (Cu-NP) on the enchytraeid Enchytraeus albidus is compared to the toxicity of a copper-salt (CuCl(2)). The effect parameters studied were
Landon John G Edgar et al.
Organic letters, 14(16), 4226-4229 (2012-08-01)
Glycosyl 1-phosphates enriched in the α-anomer are obtained without the use of protecting groups in two steps starting from the free hemiacetal. Condensation of free hemiacetals with toluenesulfonylhydrazide yields a range of glycosylsulfonohydrazide donors which can be oxidized using cupric
Yoshiaki Miyamoto et al.
Chemistry, an Asian journal, 8(4), 824-831 (2013-02-05)
The stereoselective direct transformation of N-(propargylic)hydroxylamines into cis-2-acylaziridines was achieved by the combined use of AgBF4 and CuCl. Copper salts were found to promote the transformation of the intermediary 4-isoxazolines into 2-acylaziridines and both 3-aryl- and 3-alkyl-substituted 2-acylaziridines could be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service