Skip to Content
Merck
All Photos(1)

Key Documents

05-4020

Sigma-Aldrich

Chlorosulfonic acid

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClSO3H
CAS Number:
Molecular Weight:
116.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

vapor density

4 (vs air)

vapor pressure

1 mmHg ( 25 °C)
3.3 mmHg ( 37.7 °C)

form

liquid

expl. lim.

37.7 %

availability

available only in Japan

refractive index

n20/D 1.433 (lit.)

pKa 

-6

bp

151-152 °C/755 mmHg (lit.)

density

1.753 g/mL at 25 °C (lit.)

SMILES string

OS(Cl)(=O)=O

InChI

1S/ClHO3S/c1-5(2,3)4/h(H,2,3,4)

InChI key

XTHPWXDJESJLNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guido Sonsmann et al.
Journal of the American Society for Mass Spectrometry, 13(1), 47-58 (2002-01-05)
The fragmentation of the multiply charged peptides b-chain of bovine insulin and glucagon have been investigated under low energy collision induced dissociation (CID) conditions using an electrospray ion trap mass spectrometer. The influence of charge state, specific amino acids such
P Taddei et al.
Biomacromolecules, 8(4), 1200-1208 (2007-03-07)
Silk fabrics were treated with chlorosulphonic acid in pyridine for different times. The amount of sulfur bound to silk increased during the first 2 h of reaction and then reached a plateau. The amino acidic pattern of sulfated silk remained
Yongzhen Tao et al.
Carbohydrate research, 341(13), 2261-2269 (2006-06-30)
A water-soluble hyperbranched beta-glucan, coded as TM3b, extracted from sclerotia of an edible fungus (Pleurotus tuber-regium) was fractioned into eight fractions coded as F1-F8 by a nonsolvent addition method. Five fractions were treated with chlorosulfonic acid at 35 degrees C
José Barluenga et al.
Journal of the American Chemical Society, 131(11), 4031-4041 (2009-02-28)
A detailed study of the scope of a new Pd-catalyzed synthesis of indoles from 1,2-dihaloarenes and o-halobenzene sulfonates and imines is described. The cascade reaction comprises an imine alpha-arylation followed by an intramolecular C-N bond-forming reaction promoted by the same
Yasushi Tamada
Biomaterials, 25(3), 377-383 (2003-10-31)
Silk fibroin (Bombyx mori) was sulfated using chlorosulfonic acid in pyridine. FT-IR spectra showed introduction of sulfate group by this reaction; NMR spectra indicated that sulfation occurred mainly at tyrosine and serine residues. Molecular size decreased and dispersed with sulfation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service