Skip to Content
Merck
All Photos(1)

Key Documents

05-0975

Sigma-Aldrich

Hydroxyapatite

SAJ first grade, 98.0-103.0%

Synonym(s):

Calcium phosphate hydroxide, Durapatite, Hydroxylapatite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3Ca3(PO4)2 · Ca(OH)2
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
12352305
PubChem Substance ID:

grade

SAJ first grade

Assay

98.0-103.0%

form

powder

availability

available only in Japan

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bjarke Viberg et al.
Acta orthopaedica, 84(3), 254-259 (2013-04-19)
Elderly patients with displaced femoral neck fractures are commonly treated with a hemiarthroplasty (HA), but little is known about the long-term failure of this treatment. We compared reoperation rates for patients aged at least 75 years with displaced femoral neck
Sophie E P New et al.
Circulation research, 113(1), 72-77 (2013-04-26)
We previously showed that early calcification of atherosclerotic plaques associates with macrophage accumulation. Chronic renal disease and mineral imbalance accelerate calcification and the subsequent release of matrix vesicles (MVs), precursors of microcalcification. We tested the hypothesis that macrophage-derived MVs contribute
H S Alghamdi et al.
Journal of dental research, 92(11), 982-988 (2013-09-24)
Osteoporotic conditions are anticipated to affect the osseointegration of dental implants. This study aimed to evaluate the effect of a radiofrequent magnetron-sputtered calcium phosphate (CaP) coating on dental implant integration upon installment in the femoral condyles of both healthy and
Ehud Segal et al.
PloS one, 4(4), e5233-e5233 (2009-04-22)
There is an immense clinical need for novel therapeutics for the treatment of angiogenesis-dependent calcified neoplasms such as osteosarcomas and bone metastases. We developed a new therapeutic strategy to target bone metastases and calcified neoplasms using combined polymer-bound angiogenesis inhibitors.
Sören Toksvig-Larsen et al.
Acta orthopaedica, 84(3), 314-318 (2013-04-30)
External fixation pins tend to loosen with time, especially from cancellous bone. A coating that releases a bisphosphonate has been shown to improve the fixation of dental implants in humans. We now tested a bisphosphonate coating on steel pins for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service