Skip to Content
Merck
All Photos(1)

Key Documents

05-0540

Sigma-Aldrich

Calcium chloride

≥75%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥75%

form

crystalline

availability

available only in Japan

mp

772 °C (lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Philippe et al.
International journal of obesity (2005), 39(2), 295-302 (2014-06-04)
A significant proportion of severe familial forms of obesity remain genetically elusive. Taking advantage of our unique cohort of multigenerational obese families, we aimed to assess the contribution of rare mutations in 29 common obesity-associated genes to familial obesity, and
Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
Mohsen Akbari et al.
Advanced functional materials, 24(26), 4060-4067 (2014-11-21)
The fabrication of cell-laden structures with anisotropic mechanical properties while having a precise control over the distribution of different cell types within the constructs is important for many tissue engineering applications. Automated textile technologies for making fabrics allow simultaneous control
Ilie-Cosmin Stancu et al.
Acta neuropathologica, 129(6), 875-894 (2015-04-12)
Prion-like seeding and propagation of Tau-pathology have been demonstrated experimentally and may underlie the stereotyped progression of neurodegenerative Tauopathies. However, the involvement of templated misfolding of Tau in neuronal network dysfunction and behavioral outcomes remains to be explored in detail.
Michael A Goren et al.
Nature communications, 5, 5115-5115 (2014-10-09)
Opsin, the rhodopsin apoprotein, was recently shown to be an ATP-independent flippase (or scramblase) that equilibrates phospholipids across photoreceptor disc membranes in mammalian retina, a process required for disc homoeostasis. Here we show that scrambling is a constitutive activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service