Skip to Content
Merck
All Photos(1)

Key Documents

03-3390

Sigma-Aldrich

Bromine water

CP

Synonym(s):

Bromine solution, Bromide bromate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

form

liquid

availability

available only in Japan

density

1.307 g/mL at 25 °C (lit.)

storage temp.

15-25°C

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Zhu et al.
Waste management (New York, N.Y.), 33(2), 484-488 (2012-11-28)
Separation of waste printed circuit boards (WPCBs) has been a bottleneck in WPCBs resource processing. In this study, the separation of WPCBs was performed using dimethyl sulfoxide (DMSO) as a solvent. Various parameters, which included solid to liquid ratio, temperature
José A B Ferreira et al.
The journal of physical chemistry. A, 116(48), 11938-11945 (2012-11-09)
Nucleophilic addition of water and of methanol to 3,6-diamino-2,4,5,7-tetrabromo-9-[2-(methoxycarbonyl) phenyl]-9H-xanthen-9-ylium, 4BrR123, yields respectively 2-(3,6-diamino-2,4,5,7-tetrabromo-9-hydroxy-9H-xanthen-9-yl)xanthyl benzoate, HO4BrR123 and 2-(3,6-diamino-2,4,5,7-tetrabromo-9-methoxy-9H-xanthen-9-yl)xanthyl benzoate, MeO4BrR123. The novel experimental results are addressed theoretically. The linear free energy relationship, LFER, second-order perturbation theory analysis of the natural
Chemistry. Unconventional chemistry for unconventional natural gas.
Eric McFarland
Science (New York, N.Y.), 338(6105), 340-342 (2012-10-23)
Nickel-butadiene catalytic system for the cross-coupling of bromoalkanoic acids with alkyl Grignard reagents: a practical and versatile method for preparing fatty acids.
Takanori Iwasaki et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(9), 2956-2960 (2013-02-02)
Fátima García et al.
Organic & biomolecular chemistry, 11(5), 765-772 (2012-12-18)
Bis(triazole)benzamide 1 has been readily synthesized by means of Cu-catalyzed 1,3-dipolar cycloaddition and its ability to bind halide anions and neutral gallic acid derivative 12GA has been theoretically and experimentally investigated. The cavity defined by the N-H amide group and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service