Skip to Content
Merck
All Photos(1)

Documents

03-0910

Sigma-Aldrich

Benzene

suitable for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Beilstein:
969212
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

vapor density

2.77 (vs air)

vapor pressure

166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

autoignition temp.

1043 °F

expl. lim.

8 %

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.501 (lit.)

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelle Vlaanderen et al.
Environmental health perspectives, 119(2), 159-167 (2010-10-01)
The use of occupational cohort studies to assess the association of benzene and lymphoma is complicated by problems with exposure misclassification, outcome classification, and low statistical power. We performed meta-analyses of occupational cohort studies for five different lymphoma categories: Hodgkin
Cliona M McHale et al.
Carcinogenesis, 33(2), 240-252 (2011-12-15)
Benzene causes acute myeloid leukemia and probably other hematological malignancies. As benzene also causes hematotoxicity even in workers exposed to levels below the US permissible occupational exposure limit of 1 part per million, further assessment of the health risks associated
John C Thomas et al.
ACS nano, 9(5), 4734-4742 (2015-04-14)
Carboranethiol molecules self-assemble into upright molecular monolayers on Au{111} with aligned dipoles in two dimensions. The positions and offsets of each molecule's geometric apex and local dipole moment are measured and correlated with sub-Ångström precision. Juxtaposing simultaneously acquired images, we
Deborah C Glass et al.
Occupational and environmental medicine, 71(4), 266-274 (2014-02-18)
Benzene exposure has been associated with increased risk of leukaemia and myelodysplastic syndrome. Existing studies are sparse for other lymphohaematopoietic cancer subtypes, such as myeloproliferative disease (MPD) and the related chronic myeloid leukaemia (CML). We pooled data from three petroleum
David Galbraith et al.
Critical reviews in toxicology, 40 Suppl 2, 1-46 (2010-10-14)
Over the last century, benzene has been a well-studied chemical, with some acute and chronic exposures being directly associated with observed hematologic effects in humans and animals. Chronic heavy exposures to benzene have also been associated with acute myelogenous leukemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service