Skip to Content
Merck
All Photos(1)

Key Documents

03-0670

Sigma-Aldrich

Beeswax

white, CP

Synonym(s):

Cera alba

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12181502

grade

CP

form

solid

availability

available only in Japan

color

white

mp

61-65 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiep Minh Nguyen et al.
Pest management science, 68(7), 1062-1068 (2012-03-03)
In the natural environment, photodegradation is one of the most common degradative processes of pesticides. In order to reduce the photodegradation of pesticides, and so increase their killing activity against target pests, chitosan-coated beeswax solid lipid nanoparticles (CH-BSLNs) were prepared
M Cristina Gamberini et al.
Analytical and bioanalytical chemistry, 401(6), 1839-1846 (2011-07-05)
A study of the composition of the remains of ancient ointments from museums was undertaken to enable understanding of the preparation techniques. Comparison of ancient recipes from different historical periods and spectroscopic characteristics of inorganic and/or organic remains recovered in
Randa Latif et al.
Journal of microencapsulation, 28(3), 159-165 (2011-03-24)
Octyl-p-methoxycinnamate (OMC) is a sun-blocking agent that absorbs ultraviolet (UV) radiation in UVB range. However, when exposed to sunlight, OMC is converted into a less UV-absorbent form, which reduces its effectiveness. The aim of this study was to stabilize the
Christian W W Pirk et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 181(3), 353-359 (2010-11-17)
Colonies of Apis florea, which only abscond a short distance, usually return to salvage old nest wax; but, those colonies, and all other honeybee species which go considerably further, do not. Wax salvage would clearly be counter-productive unless the energy
Peter A Lewis et al.
Journal of burn care & research : official publication of the American Burn Association, 33(4), e195-e200 (2012-06-06)
Postburn itch is reported to affect up to 87% of the burn population. Although treatments for postburn itch are multimodal, they remain consistently ineffective. However, recent anecdotal evidence from several outpatients at a tertiary referral hospital suggests that a cream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service