Skip to Content
Merck
All Photos(2)

Documents

THSNTKB

Roche

KAPA Taq HotStart

suitable for PCR, with dNTPs

Synonym(s):

PCR, hotstart PCR, taq

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 1000 reactions
sufficient for 500 reactions

shelf life

≤18 mo.

feature

dNTPs included
hotstart

packaging

pkg of 250 U (KK1509)
pkg of 500 U (KK1511)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA Taq DNA Polymerase is the single-subunit Taq DNA polymerase of the thermophilic bacterium Thermus aquaticus, purified from recombinant Escherichia coli. KAPA Taq DNA Polymerase has 5’g3’ polymerase and 5’g3’ exonuclease activity, but no 3’g5’ exonuclease (proofreading) activity. The enzyme system has an error rate of approximately 1 error per 2.2 x 105 nucleotides incorporated. In the HotStart formulation, the enzyme is combined with a proprietary antibody that inactivates the enzyme until the first denaturation step. This prevents nonspecific amplification during reaction setup, increases sensitivity, and improves reaction efficiency. PCR products generated with KAPA Taq HotStart are A-tailed and may be cloned into TA cloning vectors. KAPA Taq HotStart Buffer is a uniquely formulated buffer to facilitate specific primer annealing. This translates to higher yields of specific product when compared to traditional Taq buffers, and improved amplification of GC- and AT-rich templates. However, KAPA Taq HotStart DNA Polymerase may be used in combination with any standard Taq buffer with a pH of 8.3 or higher.

Application

KAPA Taq HotStart® has been used for:
  • High throughput PCR
  • Amplification of low copy DNA templates
  • Multiplex PCR
  • Specific amplification of complex templates
  • RT-PCR
  • Aquarium DNA extraction to generate marker gene libraries

Biochem/physiol Actions

KAPA Taq HotStart® DNA polymerase and KAPA Taq exhibit 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity. The enzyme shows an error rate of approximately 1 error per 2.2 x 105 nucleotides incorporated. In the hot start formulation, the KAPA Taq is combined with a proprietary antibody that inactivates the enzyme until the first denaturation step, eliminating spurious amplification products and increasing reaction efficiency and sensitivity. KAPA Taq hotstart buffer is formulated uniquely to facilitate specific primer annealing. This translates to higher yields of the specific product when compared to traditional Taq buffers. It also enhances the amplification of GC- and AT-rich templates. KAPA Taq hotstart DNA polymerase might be used in combination with any standard Taq buffer with a pH of 8.3 or higher

Features and Benefits

High performance
  • Improved sensitivity, specificity, and yields.
  • Novel buffer formulation facilitates specific primer annealing, leading to higher yield of specific product.

Quick Notes:
  • KAPA Taq HotStart® DNA Polymerase can replace any commercial hotstart Taq DNA polymerase in an existing protocol. The final MgCl2 concentration and annealing temperature may need to be optimized to account for differences in formulation.
  • The KAPA Taq HotStart Buffer is a uniquelyformulated buffer offering improved specificity and sensitivity, and improved amplification of GC- and AT-rich templates.
  • The KAPA Taq HotStart Buffer does not contain MgCl2; MgCl2 (25 mM) is supplied separately to allow greater flexibility during reaction setup.
  • The KAPA Taq HotStart PCR Kit is suitable for the amplification of fragments up to 3.5 kb from genomic DNA or 5 kb from less complex targets.

Quality

Each batch of KAPA Taq HotStart DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA Taq HotStart PCR kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activity, and meet strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long term storage.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.

Kit Components Only

Product No.
Description

  • KAPA Taq HotStart® DNA Polymerase 5 U/µL

  • 5X KAPA Taq HotStart® Buffer

  • MgCl2 25 mM

  • KAPA dNTP Mix 10 mM each

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holly M Bik et al.
mSphere, 4(1) (2019-02-23)
Although aquaria are common features of homes and other buildings, little is known about how environmental perturbations (i.e., tank cleaning, water changes, addition of habitat features) impact the diversity and succession of aquarium microbial communities. In this study, we sought
Important Metabolic Pathways and Biological Processes Expressed by Chicken Cecal Microbiota
Ondrej Polansky
Applied and Environmental Microbiology, 82-5, 1569-1576 (2016)

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service