LRHSRMKB
Roche
KAPA LongRange
HotStart ReadyMix™, suitable for PCR, with dye
Synonym(s):
PCR
Select a Size
$334.00
Select a Size
About This Item
$334.00
Recommended Products
Quality Level
usage
sufficient for 100 reactions
sufficient for 500 reactions
shelf life
≤18 mo.
feature
Long Range PCR
dNTPs included
hotstart
packaging
pkg of 1.25 mL (100 x 25 μL rxn; KK3601)
pkg of 6.25 mL (500 x 25 μL rxn; KK3602)
manufacturer/tradename
Roche
technique(s)
PCR: suitable
input
purified DNA
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
KAPA HotStart ReadyMix with dye Long Range is also supplied in 2X ReadyMix format with loading dye that contains all the components required for PCR except primers and template. The loading dye in the reaction buffer allows you to load PCR products directly onto the agarose gel with no extra steps.
Application
Biochem/physiol Actions
Features and Benefits
- Amplification of targets with high sensitivity and specificity up to 15 kb
- High yields with low input DNA
Outperforms with complex targets:
- Exhibits high yield, greater sensitivity and specificity where competitor long-range systems fail
Quick Notes:
- KAPA LongRange HotStart is a blend of KAPA Taq HotStart and a proofreading DNA polymerase.
- Suitable for short-, medium- and long-range PCR, and PCR with low amounts of template DNA.
- Fidelity is 2–4 times better than Taq.
- For short amplicons, replace Taq DNA polymerase for improvements in yield and/or fidelity.
- Amplify mid- to long-range targets with high yield and sensitivity.
- 2X ReadyMix with dye contains two inert tracking dyes to allow loading of PCR products directly onto agarose gels for analysis.
Quality
Preparation Note
Other Notes
Legal Information
Kit Components Only
- KAPA Long Range DNA Polymerase or KAPA Long Range HotStart® DNA Polymerase 5 U/μL or 2.5 U/μL
- 1X KAPA Long Range Buffer
- dNTP Mix 0.3mM of each dNTP at 1X
- 2X KAPA LongRange HotStart® ReadyMix with dye
- MgCl2 at 1X 2 mM
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service