Skip to Content
Merck
All Photos(1)

Key Documents

HKG6PDH-RO

Roche

Hexokinase/Glucose-6-Phosphate Dehydrogenase (HK/G6P-DH)

from yeast/Leuconostoc, overproducer

Synonym(s):

G6P-DH, Glucose-6-Phosphate DehydrogenaseHK, Hexokinase

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$360.00
10 ML
$573.00

$360.00


Available to ship on24 April 2025Details



Select a Size

Change View
5 ML
$360.00
10 ML
$573.00

About This Item

UNSPSC Code:
12352204

$360.00


Available to ship on24 April 2025Details


biological source

yeast

Quality Level

form

suspension

mol wt

dimer 110,000 Da

packaging

pkg of 10 mL (10737275001 [30 mg])
pkg of 5 mL (10127825001 [15 mg])

manufacturer/tradename

Roche

concentration

3.21 mg/mL

technique(s)

activity assay: suitable

color

white

pH

5.5-6.5

solubility

water: miscible

suitability

suitable for UV spectrophotometry and general use

application(s)

life science and biopharma

storage temp.

2-8°C

General description

Hexokinase is responsible for catalyzing the first step of glycolysis. It results in the formation of glucose-6-phosphate and ADP from glucose and ATP.[1][2] Glucose-6-phosphate dehydrogenase is a crucial enzyme in the pentose phosphate pathway. It is responsible for the conversion of glucose-6-phosphate and NADP+ (nicotinamide adenine dinucleotide phosphate) to 6-phosphogluconolactone and NADPH.[3]

Application

Hexokinase/Glucose-6-phosphate dehydrogenase has been used for the determination of plasma glucose levels.[4]

Components

EC 2.7.1.1 & 1.1.1.49

Unit Definition

One unit (U) HK will phosphorylate 1 μmol of D-glucose in one minute at +25 °C and pH 7.6.
One unit (U) G6P-DH will oxidize 1 μmol of glucose-6-phosphate in one minute at +25 °C and pH 7.6.
The coupled assay produces 1 μmol of NADH per μmol of D-glucose phosphorylated.

Volume Activity: 340 U hexokinase/ml at +25 °C with glucose and ATP as the substrates. 170 U glucose-6-phosphate dehydrogenase/ml at +25 °C with glucose-6-phosphate as the substrate

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6. Prepared by mixing hexokinase with G6P-DH. HK:G6P-DH approximately 2:1 with regard to protein content.

Other Notes

For life science research only. Not for use in diagnostic prodcedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Gottlob et al.
Genes & development, 15(11), 1406-1418 (2001-06-08)
The serine/threonine kinase Akt/PKB is a major downstream effector of growth factor-mediated cell survival. Activated Akt, like Bcl-2 and Bcl-xL, prevents closure of a PT pore component, the voltage-dependent anion channel (VDAC); intracellular acidification; mitochondrial hyperpolarization; and the decline in
H Pelicano et al.
Oncogene, 25(34), 4633-4646 (2006-08-08)
Most cancer cells exhibit increased glycolysis and use this metabolic pathway for generation of ATP as a main source of their energy supply. This phenomenon is known as the Warburg effect and is considered as one of the most fundamental
Miguel Vázquez-Velasco et al.
Nutricion hospitalaria, 32(6), 2718-2724 (2015-12-17)
Type 2 diabetes is a very prevalent chronic disease. Among dietary factors for its prevention and treatment, interest has grown in satiating fibre (konjac glucomannan) and spirulina. Our previous studies suggest that glucomannan itself and/or in conjunction to spirulina displayed
Robert C Stanton
IUBMB life, 64(5), 362-369 (2012-03-21)
Glucose-6-phosphate dehydrogenase (G6PD) is the rate-limiting enzyme of the pentose phosphate pathway. Many scientists think that the roles and regulation of G6PD in physiology and pathophysiology have been well established as the enzyme was first identified 80 years ago. And

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service