CHROMTH-RO
Roche
Chromozym TH
Synonym(s):
thrombin substrate
Select a Size
$795.00
Select a Size
About This Item
$795.00
Recommended Products
Assay
90% (Tosyl-Gly-Pro-Arg-4-nitranilide acetate (enzymatic))
90% (enzymatic)
Quality Level
form
powder
mol wt
Mr 662.62
packaging
pkg of 100 mg (11585398001)
pkg of 20 mg (10206849001)
manufacturer/tradename
Roche
shipped in
ambient
storage temp.
20-25°C
Related Categories
General description
Application
Quality
Sequence
Preparation Note
Storage conditions (working solution): 2 to 8 °C
A substrate solution (approx. 2 mM) is stable for at least 4 weeks if stored at 2 to 8 °C.
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service