Select a Size
$340.00
Select a Size
About This Item
$340.00
Recommended Products
usage
sufficient for 200 reactions
sufficient for 500 reactions
Quality Level
shelf life
≤18 mo.
feature
Fast PCR
Multiplex PCR
dNTPs included: no
hotstart: no
packaging
pkg of 100 U (KK5020)
pkg of 250 U (KK5021)
manufacturer/tradename
Roche
technique(s)
PCR: suitable
input
purified DNA
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Features and Benefits
- Extension times as low as 1 sec per kilobase
- Broad coverage of both AT- and GC-rich targets
High speed, higher performance :
- Single copy sensitivity from complex targets
- Increased yields
Quick Notes :
- KAPA2G Fast PCR Kits contain the engineered KAPA2G Fast DNA Polymerase, developed for fast PCR.
- Use 1 sec extension time for amplicons <1 kb and 15 sec/kb for longer amplicons, and save 20–70% of total reaction time.
- No need for specialized instrumentation or PCR consumables.
- Optimized buffer system offers improved yields, specificity and sensitivity, facilitating efficient primer annealing across a wide range of primer lengths, GC contents, and melting temperatures.
- Use 0.5 U KAPA2G Fast DNA Polymerase per 25 μL reaction, or less for smaller volumes.
- For high reaction efficiency, do not exceed 25 μL reaction volumes.
- KAPA2G Buffer A contains 1.5 mM MgCl2 at 1X.
- Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.
Quality
Preparation Note
Other Notes
Kit Components Only
- KAPA2G Fast DNA Polymerase 5 U/µL
- 5X KAPA2G Buffer A
- KAPA MgCl2 25 mM
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT SE 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Fast PCR reduces reaction time, enhancing efficiency with optimized cycling parameters.
An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service