Skip to Content
Merck
All Photos(1)

Key Documents

11767291910

Roche

TUNEL Label Mix

sufficient for 30 tests, pkg of 3 × 550 μL

Synonym(s):

transferase dUTP nick end labeling, tunel

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 550 μL
$300.00

$300.00


Check Cart for Availability


Select a Size

Change View
3 X 550 μL
$300.00

About This Item

UNSPSC Code:
41105500

$300.00


Check Cart for Availability

form

solution

Quality Level

usage

sufficient for 30 tests

packaging

pkg of 3 × 550 μL

manufacturer/tradename

Roche

color

colorless

solubility

water: miscible

storage temp.

−20°C

Related Categories

General description

The nucleotide-labeling mix (TUNEL Label) contains fluorescein-dUTP and -dNTPs, both needed to perform the TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) reaction for the detection of apoptosis in situ. The nucleotide-labeling mix is used in combination with the TUNEL enzyme to prepare a TUNEL reaction mixture. This reaction mixture is used to label DNA-strand breaks for detecting and quantifying apoptotic cell death at a single-cell level in cells and tissues.

Application

TUNEL Label Mix has been used for the determination of apoptosis using TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) assay and DNA nick-end labeling by the TUNEL method.[1][2]
The nucleotide-labeling mix (TUNEL Label) contains fluorescein-dUTP and -dNTPs, both needed to perform the TUNEL reaction for the detection of apoptosis in situ. The nucleotide-labeling mix is used in combination with the TUNEL Enzyme to prepare a TUNEL reaction mixture. This reaction mixture is used to label DNA-strand breaks for detecting and quantifying apoptotic cell death at a single-cell level in cells and tissues.

Preparation Note

Working solution: TUNEL Label is used in combination with the TUNEL Enzyme to prepare the TUNEL reaction mixture.
For one test: Mix 45 μl TUNEL Label with 5 μl TUNEL Enzyme prior to use. For negative control use 50 μl/test TUNEL Label only.
Storage conditions (working solution): Note: The TUNEL reaction mixture (45 μl TUNEL Label with 5 μl TUNEL Enzyme for 1 test) should be prepared just before use, and should not be stored. Keep the TUNEL reaction mixture on ice until use.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl J Wahlin et al.
PloS one, 8(11), e79140-e79140 (2013-11-19)
Vertebrate genomes undergo epigenetic reprogramming during development and disease. Emerging evidence suggests that DNA methylation plays a key role in cell fate determination in the retina. Despite extensive studies of the programmed cell death that occurs during retinal development and
Ding Tian et al.
Cell death & disease, 11(7), 526-526 (2020-07-15)
Dysfunction of endothelial progenitor cells (EPCs) is a key factor in vascular complications of diabetes mellitus. Although the roles of microRNAs and circular RNAs in regulating cell functions have been thoroughly studied, their role in regulating autophagy and apoptosis of
Liang Liu et al.
Frontiers in oncology, 11, 648152-648152 (2021-08-13)
Glioma is the most common primary tumour of the central nervous system and is considered one of the greatest challenges for neurosurgery. Mounting evidence has shown that lncRNAs participate in various biological processes of tumours, including glioma. This study aimed
Yan Yang et al.
Neural regeneration research, 15(3), 464-472 (2019-10-02)
Mitochondrial dysfunction in neurons has been implicated in hypoxia-ischemia-induced brain injury. Although mesenchymal stem cell therapy has emerged as a novel treatment for this pathology, the mechanisms are not fully understood. To address this issue, we first co-cultured 1.5 ×
Sophie A Montandon et al.
EvoDevo, 5, 33-33 (2015-02-24)
Mammals exhibit a remarkable variety of phenotypes and comparative studies using novel model species are needed to uncover the evolutionary developmental mechanisms generating this diversity. Here, we undertake a developmental biology and numerical modeling approach to investigate the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service