Skip to Content
Merck
All Photos(1)

Key Documents

11112481001

Roche

Streptavidin-β-Gal conjugate

lyophilized (clear, colorless solution after reconstitution), pkg of 500 U

Synonym(s):

streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

lyophilized (clear, colorless solution after reconstitution)

packaging

pkg of 500 U

manufacturer/tradename

Roche

storage temp.

−20°C

General description

Streptavidin is covalently coupled to β-galactosidase from E. coli.

Application

  • Use Streptavidin-β-Gal to detect biotinylated compounds using: ELISA
  • Immunohistocytochemistry[1]
  • Western blot
  • In situ hybidization[2]

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate. The following concentrations should be taken as a guideline:
  • ELISA: 100 to 500 mU/ml
  • Immunohistocytochemistry: 250 to 500 mU/ml
  • Western blot: 250 to 500 mU/ml


Storage conditions (working solution): The reconstituted solution is stable at 2 to 8 °C for four weeks. Aliquots should be stored at -15 to -25 °C.
Note: Avoid repeated freezing and thawing.

Reconstitution

Add 1 ml double-distilled water to a final concentration of 500 U/ml.

Other Notes

For life science research only. Not for use in diagnostic procedures.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramona Trebbien et al.
Virology journal, 8, 434-434 (2011-09-10)
Pigs are considered susceptible to influenza A virus infections from different host origins because earlier studies have shown that they have receptors for both avian (sialic acid-alpha-2,3-terminal saccharides (SA-alpha-2,3)) and swine/human (SA-alpha-2,6) influenza viruses in the upper respiratory tract. Furthermore
Monica Ronco et al.
Developmental biology, 313(1), 80-92 (2007-12-07)
Our understanding of the developmental mechanisms underlying the vast diversity of arthropod appendages largely rests on the peculiar case of the dipteran Drosophila melanogaster. In this insect, homothorax (hth) and extradenticle (exd) together play a pivotal role in appendage patterning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service