03734927001
Roche
Taq DNA Polymerase, GMP Grade
≥13000 units/mg protein, >98% (SDS-PAGE), sufficient for ≤2,000 reactions, Difficult Templates/Specialty Enzymes PCR
Synonym(s):
dna amplification, pcr, polymerase, primer extension
About This Item
Recommended Products
description
5 U/βl
Quality Level
Assay
>98% (SDS-PAGE)
usage
sufficient for ≤2,000 reactions
specific activity
≥13000 units/mg protein
feature
Difficult Templates/Specialty Enzymes PCR
dNTPs included: no
hotstart: no
packaging
pkg of 1,000 U
manufacturer/tradename
Roche
parameter
72 °C optimum reaction temp.
technique(s)
PCR: suitable
input
purified DNA
optimum pH
~9 (20 °C)
shipped in
dry ice
storage temp.
−20°C (−15°C to −25°C)
General description
Application
Packaging
Unit Definition
Unit Assay: Incubation buffer for assay on activated DNA:
67 mM Tris/HCl; pH 8.3/25 °C, 5 mM MgCl2, 10 mM mercaptoethanol, 0.2% polydocanol, 0.2 mg/ml gelatine, 0.2 mM each dATP, dGTP, dTTP and 0.1 mM dCTP, pH 8.3 (25 °C).
Incubation procedure:
M13mp9ss, M13 primer (17mer) and 1 μCi [α-32P]-dCTP are incubated with suitable dilutions of Taq DNA Polymerase in 50 μl incubation buffer at 65 °C for 60 min. The amount of incorporated dNTPs is determined by trichloroacetic acid precipitation.
Volume Activity: 5 U/μl
Other Notes
Legal Information
Kit Components Only
- Taq DNA Polymerase, GMP Grade 5 U/μl
- PCR Buffer with MgCl<sub>2</sub> 10x concentrated
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service