Skip to Content
Merck
All Photos(3)

Key Documents

ZLXLSD51040

Milli-Q

Milli-Q® HXSD Water Purification System

Milli-Q® HXSD Water Purification System
1 of 1 reviewers received a sample product or took part in a promotion

Centralized pure water solution for up to 800 L/day Type 2 water; for low chlorine feed water

Synonym(s):

Milli-Q® HX 7040 SD Water Purification System (LC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104202
NACRES:
JA.13

Note: For more information on this product, including pricing & availability, please contact our Technical Service Team

AC/DC input

100 - 230 V, 50 - 60 Hz

Quality Level

product line

Milli-Q® HX

packaging

pkg of 1 unit

parameter

≤20 L/min distribution flow rate
222 kg operating weight (489.4 lb)
40 L/hr make-up flow rate
800 L/day max. usage

resistivity

>5 Ω-cm, at 25°C

H × W × D

124 cm (48.8 in.) × 54.3 cm (21.4 in.) × 94.7 cm (37.3 in.)

tank volume

140 L

total organic carbon (TOC) residue

<30 ppb

input

potable tap water

output

type 2 water (> 5 MΩ·cm)

conductivity

<0.2 μS/cm at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

The Milli-Q® HX 7040 SD system is a complete, compact, connected and fully customizable pure water solution for large laboratories requiring up to 800 L/day of Type 2 pure water. These systems allow a distribution flow of up to 20 L/min for a small and mid-size loop. This enables one Milli-Q HX 7040 SD system to reliably supply the pure water needs of a large laboratory or an entire floor of a building. Tap water is first pretreated by a Progard® cartridge to efficiently remove particles, colloids, free chlorine and hardness. Advanced reverse osmosis (RO) then removes 95-99% of ions, and 99% of all dissolved organics (>200 Da), microorganisms and particles. Patented E.R.A.® (Evolutive Reject Adjustment) technology automatically assesses feed water quality to produce a constant flow rate and optimize water recovery (between 45% and 75%). The Elix® electrodeionization (EDI) module further removes ions to typically reach resistivity >5 MΩ.cm (at 25 °C). Processed pure water is then irradiated by a bactericidal UV lamp before entering the built-in storage tank. This integrated 140 L water reservoir offers protected storage plus a distribution pump provides up to 20 L/min pure water for a loop of up to 50 m long. The systems are equipped with a large, colored touchscreen display panel to facilitate maintenance, control and access to data. These central systems are embedded with our unique MyMilli-Q Remote Care online monitoring and service capability, which operates as an extension of Milli-Q® Services. This digital service lets you securely and remotely manage your system, supporting your lab′s optimal productivity.

Application

Water produced from a Milli-Q® HX 7040 SD systems can reliably feed all your Type 2 pure water needs, including:
  • Equipment and instruments such as dishwashers, autoclaves, clinical analyzers, heating baths, environmental and humidity chambers, and histopathology stainers.
  • Taps on benches for general glassware rinsing, buffer and reagent preparation.
  • Purification systems to obtain ultrapure water (e.g. Milli-Q® IQ 7000 system).

Features and Benefits

  • Optimal combination of purification technologies for a reliable flow of high-quality Type 2 pure water.
  • Integrated 140 L reservoir and pump for protected storage and distribution up to 20 L/min.
  • Superior communications interface and modern data management capabilities.
  • Customizable range of options and accessories to meet your specific requirements.
  • MyMilli-Q remote care monitoring and service capability.

Components

  • The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
  • This catalogue number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Other Notes

Directions for Use

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering and UV
  • Application: General laboratory analysis
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes and exposed to bactericidal UV lamp. Refer to system equipment user guide section "Using the System" (available on USB key in the system box)
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

E.R.A. is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
MyMilli-Q is a trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover the importance of water quality in microbiology and its impact on reliable and reproducible results in culture media preparation. Learn about the EN ISO 11133 standard, lab water solutions, and purification technologies recommended for microbiological testing.

Related Content

Milli-Q® HX 7000 SD series purifies, stores, and distributes type 2 pure water efficiently in a compact format.

How consistent is your lab’s water quality? Variations in purified water quality may be impacting your lab’s analysis and overall efficiency of operations. See how Milli-Q® lab water systems deliver confidence in consistency.

Water for Clinical Chemistry

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Pouso Alegre, MG, Brasil
    • Reviews 2
    • Vote 1
    5 out of 5 stars.

    Equipamento ideal para laboratório

    Equipamento muito robusto de ótima qualidade, ideal para uso em laboratório.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service