Key Documents
ZLXLCLPAK
Milli-Q
RO Regeneration Tool
For use with Milli-Q® CLX/HX/HR/HXSD 7000 series water purification systems
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
Pricing and availability is not currently available. Contact Technical Service
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
The RO Regeneration Tool is a water purification accessory used with high-volume water systems with poor feed water conditions requiring additional maintenance of the reverse osmosis (RO) membrane.
Application
For high volume water systems requiring additional RO membrane maintenance with poor feed water conditions.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany