Skip to Content
Merck
All Photos(1)

Key Documents

ZFSQ11504

Milli-Q

Super-Q® and Super-Q® Plus Ultrapure High-Flow Water Systems

A large volume water system that consistently delivers ultrapure water up to 12 L/min which exceeds Type 1 ASTM/CAP/NCCLS specifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104206
NACRES:
JA.13

Note: For more information on this product, including pricing & availability, please contact our Technical Service Team

AC/DC input

120 V, 60 Hz
230 V, 50 Hz

Quality Level

product line

Super-Q®

packaging

pkg of 1 unit

parameter

10-12 L/min flow rate
5 bar pressure (75 psi)
63 kg operating weight (140 lb) (system filled with water)
8000 L/day max. usage

W × H × D

1150 mm (45.25 in.) × 800 mm (31.5 in.) × 220 mm (8.75 in.)

weight

30 kg (66 lb)shipping

input

feed water nature pure water

output

product water quality: type 1 water (18.2 MΩ·cm)

General description

Super-Q® water purification systems are modular systems designed to provide the "final polish" to water that has been pre-treated by purification methods such as reverse osmosis (RO), deionization (DI), or distillation.

Super-Q® systems can produce up to 12 L/min of consistently ultrapure water that is free of ionic contamination (resistivity 18.2 MW·cm at 25 °C), which exceeds Type 1 ASTM/CAP/NCCLS specifications. Product water is free from particulate and microbiological contamination to an absolute size level determined by the pore size of the Millipore® filter used. With the appropriate purification cartridges installed in the proper configuration, ultrapure water with total organic carbon (TOC) < 20 ppb can be achieved for organic-sensitive applications.

If you are interested in this product, please contact us.

Application

Super-Q® systems produce consistent quality Type 1/ultrapure water that is ideal for critical applications requiring large volumes of water, such as preparation of reagents, buffers, culture media, or for rinsing in fields such as microelectronics.

This water may also be used for analytical techniques such as fluorimetry[1], atomic absorption (AA) spectrophotometry, and high performance liquid chromatography (HPLC)[2][3][4][5] and ion chromatography.[3]

Features and Benefits

  • Produces consistent quality ultrapure water which exceeds Type 1 ASTM/CAP/NCCLS quality specifications.
  • In-line monitoring of water quality.
  • Cartridge selection guide to tailor product water quality:
- Milligard® Cartridge Filter, 20 in. - Removes high particle loads from feed water.
- Super-C Cartridge - Removes organics using unique dry-packed activated carbon.
- Ion-Ex Cartridge - Removes inorganic ions with high efficiency.
- Organex-Q® Cartridge - Removes trace organics.
- Durapore® Cartridge - Removes all material larger than pore size of 0.22 μm.
  • Automatic Recirculation - An integrated pump automatically recirculates water for 5 minutes every 1.5 hours to maintain high quality ultrapure water between uses.
  • Usage of high-quality purification media for low extractables, both in terms of TOC and particulate levels (resin fines).
  • Select from a full range of services, including timesaving MyMilli-Q® digital services.
  • Manufactured in an ISO 9001 and ISO 14001 certified production site.

Components

  • The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
  • This catalogue number does not include purification catridges, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us.

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
ORGANEX-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SUPER-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Super-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kim van Wijck et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(26), 2794-2801 (2011-08-25)
Many pathophysiological conditions are associated with increased gastrointestinal permeability, reflecting an elevated risk of endotoxaemia, inflammation, and sepsis. Permeability tests are increasingly used in clinical practice to obtain information on gastrointestinal functioning, but tests are often restricted to the small
Monitoring emerging chlorination by-products in drinking water using UV-absorbance and fl uorescence indexes
Roccaro P and Vagliasindi G A F
Desalination and Water Treatment, 23, 118-122 (2010)
Xinxin Xu et al.
Chemosphere, 185, 833-843 (2017-07-25)
The feasibility of UV-activated peroxymonosulfate (PMS) technology for the degradation of Tris (2-chloroethyl) phosphate (TCEP) in an aqueous solution was investigated in this study. The conditions of [PMS]0: [TCEP]0 = 20:1, T = 25 ± 2 °C and pH = 5.5 ± 0.5 cause a 94.6% removal of TCEP (1 mg L-1) after
Fingerprint Analysis of Polygonatum odoratum (Mill.) Druce Using High-Performance Liquid Chromatographic Method with Diode-Array Detection
Guang Sheng Xi
Advanced Materials Research, 781-784, 1199-1202 (2013)
Victor A Vega et al.
Journal of AOAC International, 98(3), 806-809 (2015-06-19)
A quick and effective extraction of deoxynivalenol (DON) in food samples is described. The samples were extracted using 80% acetonitrile-water solution, followed by a second extraction using an 80% methanol-water solution. The extract was then subjected to an immunoaffinity column

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service