ZFCBIXXBE
Country Box Milli-Q® IX Systems
Available only in Belgium
Synonym(s):
Country box Milli-Q® IX 7003/05/10/15 BE
About This Item
feature
Quick Reference Guide and a user manual in local language on a USB key, and a power cord adapted to the local electrical network
packaging
pkg of 1 kit
availability
available only in (Belgium)
compatibility
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service