Skip to Content
Merck
All Photos(1)

Documents

YP22

Millipore

Power cord

for mA400 Basic Power Supply, mA700 Essential Power Supply and mPAGE® Lux Curing Station

Synonym(s):

mPAGE® Lux Casting System, mPAGE® Lux Curing Station, mPAGE® Lux, gel casting device, gel casting system, electrophoresis, power cord, power cord, power plug, US power cord, US power plug, mA400, mA700, Power Supply

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105303
NACRES:
NB.22

product name

YP22 EU power cord, for mA400 Basic Power Supply, mA700 Essential Power Supply and mPAGE® Lux Curing Station

AC/DC input

125 V, 10 A

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

General description

This electrical power cord has a specific plug configuration to match electrical outlets used in North America. This cord has been validated to replace the cord supplied with the mA400 Power Supply, mA700 Power Supply and the mPAGE® Lux Curing Station.

Compatibility

Compatible with:
mA400 Basic Power Supply
mA700 Essential Power Supply
mPAGE® Lux Curing Station

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service