Key Documents
Y.11086
Prove Connect to LIMS
Unlimited License
About This Item
UNSPSC Code:
12352200
General description
In many cases it is advisable to export results and measurement data sets, e.g. to archive them or to analyze them in greater detail using a suitable software tool. Spectroquant® Prove Connect to LIMS is a desktop application to connect your Spectroquant® Prove instruments to laboratory software systems such as LIMS and ELNs. The software receives meisasurement data from Spectroquant® Prove instruments over the local network and provides it to laboratory information systems.
Spectroquant® Prove Connect runs as a background service and supports the following file formats:
−XML
−CSV
This allows an easy integration of Spectroquant® Prove instruments into an existing laboratory IT infrastructure.
This item can only be used in combination with Spectroquant® Prove spectrophotometers. A PC with Microsoft Windows 7 SP1, Windows 8, or Windows 10 and 1 GB of free disk space, 4 GB of RAM, and Network Connection (LAN) is mandatory to ensure compatibility.
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Protocols
Photometric determination of phosphate ions in water with ammonium molybdate using Spectroquant® test kits and a UV/VIS spectrophotometer.
Photometric determination of nitrite ions in soil samples by Griess method using Spectroquant® test kits and spectrophotometer.
Learn about the photometric determination of nitrate ions in soil using Spectroquant® test kits and spectrophotometer.
Photometric determination of nitrite ions in compost by Griess method using Spectroquant®
See All