Skip to Content
Merck
All Photos(1)

Key Documents

XX6200036

Millipore

Replacement Kit, XX62 000 35

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$196.00

$196.00


Check Cart for Availability


Select a Size

Change View
1 EA
$196.00

About This Item

UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24

$196.00


Check Cart for Availability

shipped in

ambient

Quality Level

General description

Use polypropylene syringes to dispense liquids into small containers and pressure-filter small volumes through filter holders of filter units with female Luer-Slip inlets. Syringes have male Luer-Slip outlets. An optional two-way vacuum valve, nylon male-to-male Luer adapter and rubber tubing with male Luer inlet converts this standard syringe into a hand vacuum pump. For sterile applications, an autoclavable stainless steel vacuum syringe with stainless steel, two-way valve is available.



Applications:

Small-Volume Liquid Filtration, Hand Pump for In-Field Filtration with Sterifil Units and Other Filter Holders

Components

includes plunger O-ring, 2 valve O-rings, valve spring, seals and balls

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service