Skip to Content
Merck
All Photos(2)

Key Documents

WX0003

Supelco

Water

OmniTrace Ultra

Synonym(s):

aa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
41104213
technique(s):
ICP: suitable
application(s):
sample preparation
bp:
100 °C (lit.)
vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

23 hPa ( 20 °C)

Quality Level

form

liquid

technique(s)

ICP: suitable

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0(-25 °C77 °F)

bp

100 °C (lit.)

mp

0 °C (lit.)

solubility

water: miscible (completely)

density

1.000 g/mL at 3.98 °C (lit.)

application(s)

sample preparation

shipped in

ambient

storage temp.

room temp

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Product is double-distilled and packed in class 100 clean room conditions into preleached LDPE bottles. Each LDPE bottle is packed in an EPS innerpack and then packed into a 1x regulated carton with a Certificate of Analysis.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

  1. What are the specs of this water? I want to use it for the trace metal analysis, ppt level

    1 answer
    1. Please see the link below to review the product specifications:
      https://www.emdmillipore.com/product/Water,EMD_CHEM-WX0003#anchor_Specifications

      The product specifications are also published in the product Certificate of Analysis. Please navigate to the ‘DOCUMENTATION’ section of the Product Detail Page to access a Certificate under ‘Certificate of Analysis’. Please see the link below to review a lot specific Certificate:
      https://www.sigmaaldrich.com/product/mm/wx0003#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service