TMS-013
Fetal Bovine Serum
US Origin, sterile-filtered, suitable for cell culture
Synonym(s):
FBS, FCS, Fetal Calf Sera, Fetal Calf Serum, Fetal bovine Sera, sera, serum
About This Item
Recommended Products
biological source
bovine fetus
Quality Level
sterility
sterile; sterile-filtered
product line
FBS Classic
form
liquid
composition
hemoglobin, ≤25 mg/dL
manufacturer/tradename
Chemicon®
origin
USA origin
technique(s)
cell culture | mammalian: suitable
impurities
mycoplasma, passes test
virus, tested negative
<10 EU/mL endotoxin
storage temp.
−20°C
General description
Application
Quality
Use for general cell culture with common cell lines.
Hemoglobin level: <25.0 mg/dL
Sterility Tested: Pass
Mycoplasma Tested: Pass
Endotoxin level: <10 EU/ml
Negative for BVD, PI3, IBR, Bovine Adeno 3, Bovine Adeno 5, BRSV, REO Virus, Rabies and Blue Tongue
Origin: United States
Storage and Stability
Analysis Note
- Endotoxin and hemoglobin tested
- Tested for the presence of bacteria, virus, and mycoplasma
- Triple filtered with 0.1 micron membrane under aseptic conditions
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.
Protocols
Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.
Related Content
Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.
Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service