Skip to Content
Merck
All Photos(2)

Key Documents

SYMBDTCC1

Millipore

Steritest® Symbio 2 Media Drain Tray Container

With Curved Outlet, Streamlines the workflow, Increases Safety

Synonym(s):

Drain tray container

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,660.00

$1,660.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
$1,660.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.51

$1,660.00


Check Cart for Availability

Request more information

material

polyphenylsulfone (PPSU) container (Drain Tray Support)

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

The Steritest® Symbio 2 Media Drain Tray Container is an accessory for the Steritest® Symbio pumps. Steritest® Symbio Pumps allow reliable, faster, and easy sterility testing in a laminar flow hood, biosafety cabinet, cleanroom, or inside an isolator. The plastic fluid collector with a curved outlet for 2 canisters drain tray facilitates the installation inside an isolator.

Application

The Steritest® Symbio 2 Media Drain Tray Container is an accessory for the Steritest® Symbio pumps for streamlining the workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service