Skip to Content
Merck
All Photos(2)

Key Documents

SYMBABC01

Millipore

Steritest® Complete Ampoule Breaker Container

Accessory for the Steritest® Ampoule Breaker

Synonym(s):

Ampoule receptacle

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$2,250.00

$2,250.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
$2,250.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.51

$2,250.00


Check Cart for Availability

Request more information

material

polyphenylsulfone (PPSU) container

Agency

JP (4.06)
USP (71)
complies with EP (2.6.1)

manufacturer/tradename

Steritest®

application(s)

filtration
pharmaceutical
sample preparation
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

Steritest® complete ampoule breaker container is the actual receptacle, including seals for the broken ampoules. It is placed inside the Steritest® ampoule breaker base. It is a part of the Steritest® glass ampoule breaker and can be used as a replacement part or as a second ampoule breaker container if many ampoules are to be tested.

Application

Steritest® glass ampoule breaker is used to safely open the glass ampoules (with or without Pre-scored necks). This accessory streamlines the workflow and increases safety during laboratory filtration and sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service