Skip to Content
Merck
All Photos(1)

Documents

SX0723

Supelco

Sodium phosphate dibasic

anhydrous, HPLC, Meets ACS Specifications

Synonym(s):

Disodium Hydrogen Phosphate, Anhydrous, Disodium Hydrogen Phosphate, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7

Quality Level

Assay

≥99.0%

form

crystalline powder

technique(s)

HPLC: suitable

impurities

≤0.01% Insoluble matter

loss

≤0.2% loss on drying

pH

8.7-9.3 (20 °C, 10 g/L in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

250 °C (decomposition)

solubility

77 g/L

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

UV absorption

λ: 280 nm Amax: ≤0.05

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous, HPLC, Meets ACS Specifications

Analysis Note

Assay99.0% minChloride (Cl)0.002% maxFormWhite crystalline powderHeavy metals (as Pb)0.001% maxInsoluble matter0.01% maxIron (Fe)0.002% maxLoss on drying0.2% maxSulfate (SO4)0.005% maxUV Absorbance (1 M, water, 280 nm)0.05AU maxpH of a 5% solution at 25°C8.7-9.3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Development and validation of a headspace SPME and GC-MS method to determine alcohol content in twenty different Kombucha tea samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service