SX0216
Soda & Lime Indicating
size 4-8 mesh, meets analytical specifications of USP/NF GR ACS
Synonym(s):
Sodasorb
About This Item
Recommended Products
Agency
meets analytical specifications of USP/NF GR ACS
form
granular
size
4-8 mesh
loss
≤7% loss on drying, 200°C
color
off-white
particle size
4-12 mesh (>=80%)
shipped in
ambient
storage temp.
room temp
General description
Application
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service