Skip to Content
Merck
All Photos(1)

Key Documents

ST1705

Sigma-Aldrich

Anti-TOMM20 Mouse mAb (4F3)

liquid, clone 4F3, Calbiochem®

Synonym(s):

Anti-Translocase of Outer Mitochondrial Membrane 20

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$727.00

$727.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 μG
$727.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$727.00


Estimated to ship on26 May 2025


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F3, monoclonal

form

liquid

does not contain

preservative

species reactivity

human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOMM20(9804)

General description

Purified mouse monoclonal antibody. Recognizes the ~12-16 kDa TOMM20 protein.
Recognizes the ~12-16 kDa TOMM20 protein in HeLa, PC12, and NIH3T3 cells and human small intestine tissue.
This Anti-TOMM20 Mouse mAb (4F3) is validated for use in ELISA, Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of TOMM20.

Immunogen

Full-length, recombinant, human TOMM20 (aa 1-146), expressed as a GST fusion protein

Application






ELISA (see coments)
Immunoblotting (1-5 g/ml)
Immunocytochemistry (10 g/ml)
Paraffin Sections (3 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
293T cells
Positive Control
HeLa cells, PC12 cells, NIH3T3 cells, Human small intestine tissue

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joe C Udeochu et al.
Nature neuroscience, 26(5), 737-750 (2023-04-25)
Pathological hallmarks of Alzheimer's disease (AD) precede clinical symptoms by years, indicating a period of cognitive resilience before the onset of dementia. Here, we report that activation of cyclic GMP-AMP synthase (cGAS) diminishes cognitive resilience by decreasing the neuronal transcriptional
Adam Z Spitz et al.
Nature communications, 12(1), 1134-1134 (2021-02-20)
The BCL-2 family protein BAX has essential activity in mitochondrial regulation of cell death. While BAX activity ensures tissue homeostasis, when dysregulated it contributes to aberrant cell death in several diseases. During cellular stress BAX is transformed from an inactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service