Skip to Content
Merck
All Photos(1)

Key Documents

ST1643

Sigma-Aldrich

Anti-PDK2 Mouse mAb (2G1)

liquid, clone 2G1, Calbiochem®

Synonym(s):

Anti-Pyruvate Dehydrogenase Kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$545.00

$545.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$545.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$545.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G1, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDK2(5164)

General description

Purified mouse monoclonal antibody. Recognizes the ~40 kDa PDK2 protein.
Recognizes the ~40 kDa PDK2 protein in U-2 OS cells.
This Anti-PDK2 Mouse mAb (2G1) is validated for use in ELISA, Immunoblotting, Paraffin Sections for the detection of PDK2.

Immunogen

A recombinant polypeptide corresponding to amino acids of 187-276 human PDK2, expressed as a GST fusion protein

Application



ELISA (see comments)
Immunoblotting (1-5 g/ml)
Paraffin Sections (0.8 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
293T cells
Positive Control
U-2 OS cells

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anders Gudiksen et al.
Physiological reports, 5(7) (2017-04-13)
The purpose of the present study was to examine whether lack of skeletal muscle peroxisome proliferator-activated receptor gamma coactivator 1 alpha (PGC-1α) affects the switch in substrate utilization from a fed to fasted state and the fasting-induced pyruvate dehydrogenase (PDH)
Anders Gudiksen et al.
American journal of physiology. Endocrinology and metabolism, 312(3), E204-E214 (2016-12-29)
Fasting prompts a metabolic shift in substrate utilization from carbohydrate to predominant fat oxidation in skeletal muscle, and pyruvate dehydrogenase (PDH) is seen as a controlling link between the competitive oxidation of carbohydrate and fat during metabolic challenges like fasting.
Jakob G Knudsen et al.
Physiological reports, 4(9) (2016-05-18)
Interleukin (IL)-6 is released from skeletal muscle (SkM) during exercise and has been shown to affect hepatic metabolism. It is, however, unknown whether SkM IL-6 is involved in the regulation of exercise training-induced counteraction of changes in carbohydrate and lipid
Anders Gudiksen et al.
PloS one, 11(6), e0156460-e0156460 (2016-06-22)
Pyruvate dehydrogenase (PDH) plays a key role in the regulation of skeletal muscle substrate utilization. IL-6 is produced in skeletal muscle during exercise in a duration dependent manner and has been reported to increase whole body fatty acid oxidation, muscle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service